accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 693-65-2

IUPAC Name: 1-(pentyloxy)pentane
Synonyms: di-N-pentylether
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: AOPDRZXCEAKHHW-UHFFFAOYSA-N
SMILES: CCCCCOCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results