accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CCH3H3CNH2HOHCl

CAS RN 69320-90-7

IUPAC Name: tert-butyl 2-amino-3-hydroxybutanoate hydrochloride
Synonyms: tert-butyl 2-amino-3-hydroxybutanoate hydrochloride
Molecular Weight (g/mol): 211.69
Molecular Formula: C8H18ClNO3
InChi Key: OWBFRQWDQDYNNF-UHFFFAOYNA-N
SMILES: Cl.CC(O)C(N)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results