Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Threonine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. T24355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24355G 5 g
T24351G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24355G Supplier TCI America Supplier No. T24355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69320-90-7
Molecular Formula C8H18ClNO3
Molecular Weight (g/mol) 211.686
MDL Number MFCD00672361
InChI Key OWBFRQWDQDYNNF-IBTYICNHSA-N
Synonym H-Thr-OtBu.HCl
PubChem CID 44629865
IUPAC Name tert-butyl (2S,3R)-2-amino-3-hydroxybutanoate;hydrochloride
SMILES CC(C(C(=O)OC(C)(C)C)N)O.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 211.69
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Threonine tert-Butyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.