accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFClN

CAS RN 693245-52-2

IUPAC Name: 3-chloro-5-(trifluoromethyl)benzonitrile
Synonyms: 3-chloro-5-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 205.56
Molecular Formula: C8H3ClF3N
InChi Key: RLHYYDKFOQSWIB-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Cl)=CC(=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results