Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-(trifluoromethyl)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. C33075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33075G 5 g
C330725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33075G Supplier TCI America Supplier No. C33075G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693245-52-2
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.56
MDL Number MFCD04115854
InChI Key RLHYYDKFOQSWIB-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl benzonitrile, 5-chloro-3-cyanobenzotrifluoride, 3-chloro-5-trifluoromethyl-benzonitrile, 3-chloro-5-cyanobenzotrifluoride, benzonitrile, 3-chloro-5-trifluoromethyl, 5-chloro-3-cyano benzotrifluoride, pubchem16810, 5-chloro-3-trifluoromethyl benzenecarbonitrile
PubChem CID 4584933
IUPAC Name 3-chloro-5-(trifluoromethyl)benzonitrile
SMILES FC(F)(F)C1=CC(Cl)=CC(=C1)C#N

Specifications

Color Colorless
Quantity 5 g
UN Number 3276
Formula Weight 205.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.