accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 69367-32-4

IUPAC Name: 4'-(decyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Synonyms: 4'-(decyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Molecular Weight (g/mol): 354.49
Molecular Formula: C23H30O3
InChi Key: HQVTYOXUVQQMLD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC1=CC=C(C=C1)C1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results