Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Decyloxybiphenyl-4-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D20411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20411G Supplier TCI America Supplier No. D20411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69367-32-4
Molecular Formula C23H30O3
Molecular Weight (g/mol) 354.49
MDL Number MFCD00130126
InChI Key HQVTYOXUVQQMLD-UHFFFAOYSA-N
PubChem CID 11451085
IUPAC Name 4-(4-decoxyphenyl)benzoic acid
SMILES CCCCCCCCCCOC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 354.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Decyloxybiphenyl-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.