accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6938-06-3

IUPAC Name: butyl pyridine-3-carboxylate
Synonyms: nicotinic acid, butyl ester
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: DQULIMIQTCDUAN-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results