Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyl Nicotinate 98.0+%, TCI America™
SDP

Catalog No. N008425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N008425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N008425G Supplier TCI America Supplier No. N008425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity laboratory reagent with a minimum purity of 98.0% by GC.

Chemical Identifiers

CAS 6938-06-3
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00006390
InChI Key DQULIMIQTCDUAN-UHFFFAOYSA-N
Synonym Nicotinic Acid Butyl Ester
PubChem CID 81353
IUPAC Name butyl pyridine-3-carboxylate
SMILES CCCCOC(=O)C1=CN=CC=C1

Specifications

Color Yellow
Boiling Point 252°C
Quantity 25 g
Formula Weight 179.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Butyl Nicotinate
RTECSNumber QT0875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.