accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6939-75-9

IUPAC Name: benzyl 4-oxopentanoate
Synonyms: benzyl 4-oxopentanoate
Molecular Weight (g/mol): 206.24
Molecular Formula: C12H14O3
InChi Key: KWQUVANYFZOCEA-UHFFFAOYSA-N
SMILES: CC(=O)CCC(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results