Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Levulinate 96%, Thermo Scientific™

Catalog No. AC250162500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC250162500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC250162500 Supplier Thermo Scientific Chemicals Supplier No. 250162500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6939-75-9
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
InChI Key KWQUVANYFZOCEA-UHFFFAOYSA-N
Synonym benzyl levulinate, pentanoic acid, 4-oxo-, phenylmethyl ester, unii-am34x4q295, benzyl 4-oxovalerate, levulinate benzyl ester, phenethyl levulinate, phenylmethyl 4-oxopentanoate, 4-oxopentanoic acid phenylmethyl ester
PubChem CID 81359
IUPAC Name benzyl 4-oxopentanoate
SMILES CC(=O)CCC(=O)OCC1=CC=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.5060 to 1.5090 (20°C, 589nm)
Quantity 250 g
Formula Weight 206.22
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material Benzyl levulinate, 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.