accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOSOOONH4NH4

CAS RN 6939-89-5

IUPAC Name: diammonium 2-sulfonatobenzoate
Synonyms: diammonium 2-sulfonatobenzoate
Molecular Weight (g/mol): 236.24
Molecular Formula: C7H12N2O5S
InChi Key: HYTQXUAMMSMMBW-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[O-]C(=O)C1=CC=CC=C1S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results