Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Sulfobenzoic Acid Monoammonium Salt 98.0+%, TCI America™
SDP

Catalog No. S012525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S012525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012525G Supplier TCI America Supplier No. S012525G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6939-89-5
Molecular Formula C7H12N2O5S
Molecular Weight (g/mol) 236.24
MDL Number MFCD00040590
InChI Key HYTQXUAMMSMMBW-UHFFFAOYSA-N
PubChem CID 53442832
IUPAC Name diammonium 2-sulfonatobenzoate
SMILES [NH4+].[NH4+].[O-]C(=O)C1=CC=CC=C1S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 219.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Sulfobenzoic Acid Monoammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.