accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 694-32-6

IUPAC Name: 1-methylimidazolidin-2-one
Synonyms: 1-methylimidazolidin-2-one
Molecular Weight (g/mol): 100.12
Molecular Formula: C4H8N2O
InChi Key: JTPZTKBRUCILQD-UHFFFAOYSA-N
SMILES: CN1CCNC1=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)