accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 694-48-4

IUPAC Name: 1,3-dimethyl-1H-pyrazole
Synonyms: 1,3-dimethylpyrazole
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: NODLZCJDRXTSJO-UHFFFAOYSA-N
SMILES: CN1C=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results