Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylpyrazole 98.0+%, TCI America™
SDP

Catalog No. D45285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45285G Supplier TCI America Supplier No. D45285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 694-48-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.133
MDL Number MFCD00036665
InChI Key NODLZCJDRXTSJO-UHFFFAOYSA-N
PubChem CID 79096
IUPAC Name 1,3-dimethylpyrazole
SMILES CC1=NN(C=C1)C

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 1993
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.