accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 694-72-4

IUPAC Name: octahydropentalene
Synonyms: bicyclo[3.3.0]octane
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: AEBWATHAIVJLTA-UHFFFAOYSA-N
SMILES: C1CC2CCCC2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)