Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[3.3.0]octane, Thermo Scientific™

Catalog No. AC468522500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468522500 250 mg
AC468520010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC468522500 Supplier Thermo Scientific Chemicals Supplier No. 468522500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 694-72-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number 00060882
InChI Key AEBWATHAIVJLTA-UHFFFAOYSA-N
IUPAC Name octahydropentalene
SMILES C1CC2CCCC2C1

Specifications

Alpha Vector BICYCLOOCTANE
Color Colorless
Boiling Point 137°C
Flash Point 26°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material Bicyclo[3.3.0]octane
Hazard Category Flame
Hazard Statement Exclamation mark
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.