accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 694-83-7

IUPAC Name: (1R,2S)-cyclohexane-1,2-bis(aminium)
Synonyms: (1R,2S)-cyclohexane-1,2-bis(aminium)
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: SSJXIUAHEKJCMH-OLQVQODUSA-P
SMILES: [NH3+][C@H]1CCCC[C@H]1[NH3+]
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (6)
  • (4)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (6)
  • (4)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)