Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Diaminocyclohexane, mixture of cis and trans

Catalog No. 13255150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-255-150ML 50 mL
13-255-118L 18 L
13-255-11L 1 L
13-255-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-255-150ML Supplier Sigma Aldrich Supplier No. 13255150ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2-diaminocyclohexane undergoes non-templated reaction with homochiral as well as the racemic form of trans-1,2-diaminocyclohexane with terephthaldehyde to yield (3+3)-cyclocondensed molecular triangles
  • It acts as ligand and forms organotin complexes, having potential applications as metal-based antitumour drugs.1,2-diaminocyclohexane was used in the synthesis of chiral ruthenium(iv)-oxo complexes

Specifications

CAS 694-83-7
Density 0.931 g/mL (at 25°C (literature))
Boiling Point 92°C to 93°C (18 mmHg)
Molecular Formula C6H14N2
Refractive Index n20/D 1.49 (literature)
Linear Formula C6H10(NH2)2
MDL Number MFCD00001491
Quantity 50 mL
Synonym 1,2-Cyclohexanediamine; DHC 99
Molecular Weight (g/mol) 114.19
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.