accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69444-47-9

IUPAC Name: tetrafluoroboranuide; triethyl(methyl)azanium
Synonyms: triethyl(methyl)azanium tetrafluoroborate
Molecular Weight (g/mol): 203.03
Molecular Formula: C7H18BF4N
InChi Key: OHZMHURLSZDGTO-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[N+](C)(CC)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results