Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylmethylammonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. T219825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T219825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T219825G Supplier TCI America Supplier No. T219825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 69444-47-9
Molecular Formula C7H18BF4N
Molecular Weight (g/mol) 203.032
InChI Key OHZMHURLSZDGTO-UHFFFAOYSA-N
PubChem CID 11745724
IUPAC Name triethyl(methyl)azanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[N+](C)(CC)CC

Specifications

Melting Point 320°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 203.03
Percent Purity ≥98.0% (N)
Physical Form Crystalline Lumps
Chemical Name or Material Triethylmethylammonium Tetrafluoroborate
EINECSNumber (2)-0186
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.