accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6945-67-1

IUPAC Name: 2-bromo-4-nitropyridine
Synonyms: 2-bromo-4-nitropyridine
Molecular Weight (g/mol): 203
Molecular Formula: C5H3BrN2O2
InChi Key: AFVITJKRFRRQKT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=NC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results