Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-nitropyridine 95.0+%, TCI America™
SDP

Catalog No. B35235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35235G 5 g
B35231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35235G Supplier TCI America Supplier No. B35235G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6945-67-1 | C5H3BrN2O2 | 203.00 g/mol

Chemical Identifiers

CAS 6945-67-1
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD03701383
InChI Key AFVITJKRFRRQKT-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-4-nitro, 2-brom-4-nitropyridin, pubchem13197, acmc-1badj, 2-bromo-4-nitro-pyridine, 2-bromanyl-4-nitro-pyridine, ksc494c7j, 2-bromo-4-nitropyridine, abbypharma ap-10-5158
PubChem CID 243015
IUPAC Name 2-bromo-4-nitropyridine
SMILES [O-][N+](=O)C1=CC=NC(Br)=C1

Specifications

Melting Point 62°C
Quantity 5 g
Formula Weight 203.00
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.