CAS RN 6946-10-7
IUPAC Name:
3-(acetyloxy)-2-oxopropyl acetate
Synonyms:
3-(acetyloxy)-2-oxopropyl acetate
Molecular Weight (g/mol):
174.15
Molecular Formula:
C7H10O5
InChi Key:
PZVCVSQSQHGBNE-UHFFFAOYSA-N
SMILES:
CC(=O)OCC(=O)COC(C)=O
1
–
1
of
1
results
1
1,3-Diacetoxyacetone 98.0+%, TCI America™
CAS: 6946-10-7 Molecular Formula: C7H10O5 Molecular Weight (g/mol): 174.15 MDL Number: MFCD01310655 InChI Key: PZVCVSQSQHGBNE-UHFFFAOYSA-N Synonym: 1,3-Diacetoxy-2-propanone PubChem CID: 243418 IUPAC Name: (3-acetyloxy-2-oxopropyl) acetate SMILES: CC(=O)OCC(=O)COC(=O)C