Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diacetoxyacetone 98.0+%, TCI America™
SDP

Catalog No. D28435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28435G 5 g
D284325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28435G Supplier TCI America Supplier No. D28435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6946-10-7
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD01310655
InChI Key PZVCVSQSQHGBNE-UHFFFAOYSA-N
Synonym 1,3-Diacetoxy-2-propanone
PubChem CID 243418
IUPAC Name (3-acetyloxy-2-oxopropyl) acetate
SMILES CC(=O)OCC(=O)COC(=O)C

Specifications

Melting Point 50°C
Quantity 5 g
Formula Weight 174.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diacetoxyacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.