accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3HN

CAS RN 69478-75-7

IUPAC Name: N,N-dimethylpyrrolidin-3-amine
Synonyms: N,N-dimethylpyrrolidin-3-amine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: AVAWMINJNRAQFS-UHFFFAOYNA-N
SMILES: CN(C)C1CCNC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results