Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. D185910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D185910G 10 g
D185925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D185910G Supplier TCI America Supplier No. D185910G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69478-75-7
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD01569246
InChI Key AVAWMINJNRAQFS-UHFFFAOYSA-N
PubChem CID 2758519
IUPAC Name N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 10 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.