accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOH

CAS RN 6948-88-5

IUPAC Name: 4-[hydroxy(4-hydroxynaphthalen-1-yl)phenylmethyl]naphthalen-1-ol
Synonyms: 4-[hydroxy(4-hydroxynaphthalen-1-yl)phenylmethyl]naphthalen-1-ol
Molecular Weight (g/mol): 392.45
Molecular Formula: C27H20O3
InChi Key: OUYLDJXFDLBCTQ-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(C=C1)C(O)(C1=CC=CC=C1)C1=C2C=CC=CC2=C(O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results