Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Naphtholbenzion, MP Biomedicals™

Catalog No. ICN15578525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15578525 25 g
ICN15578505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15578525 Supplier MP Biomedicals, Inc Supplier No. 0215578525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6948-88-5
Molecular Formula C27H20O3
Molecular Weight (g/mol) 392.454
InChI Key OUYLDJXFDLBCTQ-UHFFFAOYSA-N
Synonym bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
PubChem CID 81382
IUPAC Name 4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
SMILES C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O

Specifications

Color Red to Brown
Quantity 25 g
Formula Weight 392.5
Physical Form Powder
Chemical Name or Material α-Naphtholbenzion
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.