accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 695-56-7

IUPAC Name: 1,4-dioxaspiro[4.4]non-6-ene
Synonyms: 1,4-dioxaspiro[4.4]non-6-ene
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: ATFWVBMLTLPOHI-UHFFFAOYSA-N
SMILES: C1COC2(CCC=C2)O1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)