Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyclopenten-1-one Ethylene Ketal 97.0+%, TCI America™
SDP

Catalog No. C16025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16025G Supplier TCI America Supplier No. C16025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 695-56-7
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00005408
InChI Key ATFWVBMLTLPOHI-UHFFFAOYSA-N
Synonym 1,4-dioxaspiro 4.4 non-6-ene, 2-cyclopenten-1-one ethylene ketal, cyclopent-2-en-1-one-ethyleneacetal, cyclopentenone ethyleneketal, acmc-209o9c, 6,9-dioxaspiro 4.4 non-3-ene, 1,4-dioxa-spiro 4.4 non-6-ene
PubChem CID 69668
IUPAC Name 1,4-dioxaspiro[4.4]non-8-ene
SMILES C1CC2(C=C1)OCCO2

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
UN Number 1993
Formula Weight 126.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopenten-1-one Ethylene Ketal
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.