accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69541-62-4

ClClOOOHOOHN(R)CH3H3CCH3

CAS RN 69541-62-4

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-{4-[(2,6-dichlorophenyl)methoxy]phenyl}propanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-{4-[(2,6-dichlorophenyl)methoxy]phenyl}propanoic acid
Molecular Weight (g/mol): 440.32
Molecular Formula: C21H23Cl2NO5
InChi Key: DODHIGHXRDNRPP-GOSISDBHSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(OCC2=C(Cl)C=CC=C2Cl)C=C1)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)