accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3N

CAS RN 696-30-0

IUPAC Name: 4-(propan-2-yl)pyridine
Synonyms: pyridine, 4-(1-methylethyl)-
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: FRGXNJWEDDQLFH-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=NC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results