Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylpyridine 98.0+%, TCI America™
SDP

Catalog No. I01745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I01745G 5 g
I017425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I01745G Supplier TCI America Supplier No. I01745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 696-30-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00039719
InChI Key FRGXNJWEDDQLFH-UHFFFAOYSA-N
PubChem CID 69674
IUPAC Name 4-(propan-2-yl)pyridine
SMILES CC(C)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 178°C
Quantity 5 g
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.