accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NI

CAS RN 696-40-2

IUPAC Name: 1-(3-iodophenyl)methanamine
Synonyms: 1-(3-iodophenyl)methanamine
Molecular Weight (g/mol): 233.05
Molecular Formula: C7H8IN
InChi Key: LQLOGZQVKUNBRX-UHFFFAOYSA-N
SMILES: NCC1=CC=CC(I)=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)