accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3O

CAS RN 6963-44-6

IUPAC Name: 5-(acetyloxy)pentyl acetate
Synonyms: 5-(acetyloxy)pentyl acetate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: PIJBVCVBCQOWMM-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCOC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results