Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diacetoxypentane, TCI America™
SDP

Catalog No. P1238500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1238500G 500 g
P123825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1238500G Supplier TCI America Supplier No. P1238500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6963-44-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.22
MDL Number MFCD00014985
InChI Key PIJBVCVBCQOWMM-UHFFFAOYSA-N
Synonym 1,5-diacetoxypentane, 1,5-pentanediol diacetate, pentamethylene acetate, 1,5-pentanediol, diacetate, 1,5-pentanediol, 1,5-diacetate, 5-acetyloxy pentyl acetate, pentane-1,5-diyl diacetate, 1,5-diacetoxy-pentane, pentamethylene diacetate, acmc-209o9x
PubChem CID 81414
IUPAC Name 5-(acetyloxy)pentyl acetate
SMILES CC(=O)OCCCCCOC(C)=O

Specifications

Color Colorless
Boiling Point 108°C
Quantity 500 g
Formula Weight 188.22
Physical Form Liquid
Chemical Name or Material 1,5-Diacetoxypentane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.