accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHN

CAS RN 6967-12-0

IUPAC Name: 1H-indazol-6-amine
Synonyms: 6-aminoindazole
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: KEJFADGISRFLFO-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=NNC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results