Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminoindazole 98.0+%, TCI America™
SDP

Catalog No. A079825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A079825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A079825G Supplier TCI America Supplier No. A079825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6967-12-0
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005696
InChI Key KEJFADGISRFLFO-UHFFFAOYSA-N
Synonym 6-aminoindazole, 6-amino-1h-indazole, 2h-indazol-6-amine, 1h-indazol-6-ylamine, 6-amino-indazole, indazol-6-ylamine, 6-aminoimidazole, 6-aminobenzopyrazole, 1h-indazole-6-ylamine, indazol-6-amine
PubChem CID 81423
IUPAC Name 1H-indazol-6-amine
SMILES NC1=CC=C2C=NNC2=C1

Specifications

Melting Point 206°C
Quantity 25 g
Formula Weight 133.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminoindazole
RTECSNumber NK7712000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.