accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69687-80-5

IUPAC Name: methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Synonyms: methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: OQWZEJIISPYZPW-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)NC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results