Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate, 95%, Thermo Scientific™

Catalog No. SP00004EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP00004EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP00004EA Supplier Thermo Scientific Chemicals Supplier No. SP00004EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69687-80-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00203859
InChI Key OQWZEJIISPYZPW-UHFFFAOYSA-N
Synonym methyl 2,5-dimethylpyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl-, methyl ester, 2,5-dimethyl-1h-pyrrole-3-carboxylic acid methyl ester, maybridge1_008495, acmc-1b2uc, 2,5-dimethyl,4-carbomethoxy pyrrole, 2,5-dimethylpyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylicacid,2,5-dimethyl-,methyl ester, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl-, methyl ester
PubChem CID 592729
IUPAC Name methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
SMILES CC1=CC(=C(N1)C)C(=O)OC

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 153.18
Percent Purity 95%
Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.