accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6969-16-0

IUPAC Name: (2Z)-tridec-2-enoic acid
Synonyms: (2Z)-tridec-2-enoic acid
Molecular Weight (g/mol): 212.33
Molecular Formula: C13H24O2
InChi Key: GQVYBECSNBLQJV-QXMHVHEDSA-N
SMILES: CCCCCCCCCC\C=C/C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results