Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Tridecenoic Acid 80.0+%, TCI America™
SDP

Catalog No. T07595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T07595G 5 g
T075925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T07595G Supplier TCI America Supplier No. T07595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6969-16-0
Molecular Formula C13H24O2
Molecular Weight (g/mol) 212.33
MDL Number MFCD00020487
InChI Key GQVYBECSNBLQJV-QXMHVHEDSA-N
PubChem CID 5282735
IUPAC Name (2Z)-tridec-2-enoic acid
SMILES CCCCCCCCCC\C=C/C(O)=O

Specifications

Quantity 5 g
Formula Weight 212.33
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Tridecenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.