accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HO

CAS RN 697-37-0

IUPAC Name: 1-(prop-1-yn-1-yl)cyclohexan-1-ol
Synonyms: 1-(prop-1-yn-1-yl)cyclohexan-1-ol
Molecular Weight (g/mol): 138.21
Molecular Formula: C9H14O
InChi Key: UTISCLIDFORRJP-UHFFFAOYSA-N
SMILES: CC#CC1(O)CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results