Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Propynyl)cyclohexanol, 98%, Thermo Scientific™

Catalog No. AAB2203818
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2203818 50 g
AAB2203809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2203818 Supplier Thermo Scientific Chemicals Supplier No. B2203818

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-37-0
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
MDL Number MFCD00078311
InChI Key UTISCLIDFORRJP-UHFFFAOYSA-N
Synonym 1-1-propynyl cyclohexanol, 1-prop-1-yn-1-yl cyclohexan-1-ol, 1-prop-1-yn-1-yl cyclohexanol, 1-1-propynyl cyclohexanol 10g
PubChem CID 230341
IUPAC Name 1-prop-1-ynylcyclohexan-1-ol
SMILES CC#CC1(CCCCC1)O

Specifications

Melting Point 46°C to 50°C
Boiling Point 91°C to 92°C (10 mmHg)
Quantity 50 g
Beilstein 1856828
Formula Weight 138.21
Percent Purity 98%
Chemical Name or Material 1-(1-Propynyl)cyclohexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.