accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 697-82-5

IUPAC Name: 2,3,5-trimethylphenol
Synonyms: 2,3,5-trimethylphenol isopseudocumenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: OGRAOKJKVGDSFR-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(O)=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)