accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6971-45-5

IUPAC Name: (2-methoxyphenyl)hydrazine hydrochloridyl
Synonyms: (2-methoxyphenyl)hydrazine hydrochloridyl
Molecular Weight (g/mol): 173.62
Molecular Formula: C7H10ClN2O
InChi Key: GWTHJHXBXQUILT-UHFFFAOYSA-N
SMILES: [Cl].COC1=CC=CC=C1NN
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results