Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methoxyphenylhydrazine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M06175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06175G 5 g
M061725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06175G Supplier TCI America Supplier No. M06175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6971-45-5
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
MDL Number MFCD00035456
InChI Key HECSHXUNOVTAIJ-UHFFFAOYSA-N
Synonym 2-Hydrazinoanisole Hydrochloride
PubChem CID 2849445
IUPAC Name (2-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=CC=C1NN.Cl

Specifications

Quantity 5 g
Formula Weight 174.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxyphenylhydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.