accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONH3COCl

CAS RN 6972-27-6

IUPAC Name: 6-chloro-1,3-dimethyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 6-chloro-1,3-dimethylpyrimidine-2,4-dione
Molecular Weight (g/mol): 174.58
Molecular Formula: C6H7ClN2O2
InChi Key: VATQPUHLFQHDBD-UHFFFAOYSA-N
SMILES: CN1C(Cl)=CC(=O)N(C)C1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)